Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bcl-XL Mouse anti-Mouse, Clone: 7D9, MilliporeSigma™

Catalog No. MAB4619MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4619MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4619MI Supplier MilliporeSigma Supplier No. MAB4619
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Bcl-XL Antibody, N-terminus, clone 7D9 is validated for use in flow cytometry, immunofluorescence for the detection of Bcl-XL.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bcl-XL
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone 7D9
Conjugate Unconjugated
Formulation Immunoglobulin purified by protein A affinity chromatography. Liquid in 10mM PBS, pH 7.4, with 0.2% BSA and 0.09% sodium azide.
Gene Accession No. NM_001191.2; NM_138578.1
Gene Symbols BCL2L1, BCLX, bcl-xL, DKFZp781P2092, BCL-XL/S, Bcl-X, BCL2L, bcl-xS
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 35-50 (TEAPEETEAERETPSA) of mouse Bcl-XL protein. This sequence is conserved at 15 of 16 residues in rat Bcl-XL.
Quantity 50 μg
Primary or Secondary Primary
Target Species Mouse
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.