Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Bcl-xL Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB3116
Description
Detect Bcl-xL using this Anti-Bcl-xL Antibody, Phospho-specific validated for use in western blotting.
Specifications
BCl−xL | |
Polyclonal | |
Liquid rabbit serum containing no preservatives. | |
Rabbit | |
100 μL | |
NM_001191.2; NM_138578.1 | |
−80°C for up to 12 months. Avoid repeated freeze/thaw cycles. |
Western Blot | |
Unconjugated | |
BCL2L1; DKFZp781P2092; BCl−XL/S; BCLX; bCl−xL; BCl−X; BCL2L; bCl−xS | |
Synthetic phosporylated peptide corresponding to a sequence encompassing serine 62 of human BCl−xL conjugated to KLH. | |
Primary | |
Human | |
Serum |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction