Learn More
Description
Specifications
Specifications
| Antigen | BCl−XL (pS62) |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol, with 1.0mg/mL BSA (IgG, protease free) as a carrier. 0.05% sodium azide. |
| Gene Symbols | BAD; BBC2; BBC6; BCL2L8 |
| Host Species | Rabbit |
| Immunogen | The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human BCl−xL that contains serine 62. The sequence is conserved in mouse and rat. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.