Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-cell CLL/lymphoma 11A (zinc finger protein), Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018493 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-493 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-493 Supplier Abnova Corporation Supplier No. H00053335D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BCL11A protein.

This gene encodes a C2H2 type zinc-finger protein by its similarity to the mouse Bcl11a/Evi9 protein. The corresponding mouse gene is a common site of retroviral integration in myeloid leukemia, and may function as a leukemia disease gene, in part, through its interaction with BCL6. During hematopoietic cell differentiation, this gene is down-regulated. It is possibly involved in lymphoma pathogenesis since translocations associated with B-cell malignancies also deregulates its expression. Multiple transcript variants encoding several different isoforms have been found for this gene. [provided by RefSeq

Sequence: MSRRKQGKPQHLSKREFSPEPLEAILTDDEPDHGPLGAPEGDHDLLTCGQCQMNFPLGDILIFIEHKRKQCNGSLCLEKAVDKPPSPSPIEMKKASNPVEVGIQVTPEDDDCLSTSSRGICPKQEHIADKLLHWRGLSSPRSAHGALIPTPGMSAEYAPQGICKDEPSSYTCTTCKQPFTSAWFLLQHAQNTHGLRIYLESEHGSPLTPRVLHTPPFGVVPRELKMCGSFRMEAREPLSSEKI

Specifications

Antigen B-cell CLL/lymphoma 11A (zinc finger protein)
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BCL11A protein.
Formulation PBS with no preservative; pH 7.4
Gene BCL11A
Gene Accession No. NM_138559.1
Gene Alias BCL11A-L/BCL11A-S/BCL11A-XL/CTIP1/EVI9/FLJ10173/FLJ34997/HBFQTL5/KIAA1809/ZNF856
Gene Symbols BCL11A
Host Species Rabbit
Immunogen BCL11A (NP_612569.1, 1 a.a. ∼ 243 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 53335
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.