Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-cell CLL/lymphoma 7B, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018156 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-156 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-156 Supplier Abnova Corporation Supplier No. H00009275D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human BCL7B protein.

The protein encoded by this gene contains a region that is highly similar to the N-terminal segment of BCL7A protein. The BCL7A protein is encoded by the gene known to be directly involved in a three-way gene translocation in a Burkitt lymphoma cell line. This gene is located at a chromosomal region commonly deleted in Williams syndrome. The function of this gene has not yet been determined. [provided by RefSeq

Sequence: MSGRSVRAETRSRAKDDIKKVMAAIEKVRKWEKKWVTVGDTSLRIFKWVPVTDSKEKEKSKSNSSAAREPNGFPSDASANSSLLLEFQDENSNQSSVSDVYQLKVDSSTNSSPSPQQSESLSPAHTSDFRTDDSQPPTLGQEILEEPSLPSSEVADEPPTLTKEEPVPLETQVVEEEEDSGAPPLKRFCVDQPTVPQTASES

Specifications

Antigen B-cell CLL/lymphoma 7B
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human BCL7B protein.
Formulation PBS with no preservative; pH 7.4
Gene BCL7B
Gene Accession No. NM_001707.2
Gene Symbols BCL7B
Host Species Rabbit
Immunogen BCL7B (NP_001698.2, 1 a.a. ∼ 202 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9275
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.