Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

B-cell CLL/lymphoma 9, Mouse, Clone: 2D4, Abnova™

Catalog No. 89014060 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-060 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-060 Supplier Abnova Corporation Supplier No. H00000607M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BCL9.

BCL9 is associated with B-cell acute lymphoblastic leukemia. It may be a target of translocation in B-cell malignancies with abnormalities of 1q21. Its function is unknown. The overexpression of BCL9 may be of pathogenic significance in B-cell malignancies. [provided by RefSeq

Sequence: DDDSPPARSPNLPSMNNMPGMGINTQNPRISGPNPVVPMPTLSPMGMTQPLSHSNQMPSPNAVGPNIPPHGVPMGPGLMSHNPIMGHGSQEPPMVPQGRM

Specifications

Antigen B-cell CLL/lymphoma 9
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 2D4
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BCL9.
Formulation PBS with no preservative; pH 7.4
Gene BCL9
Gene Accession No. NM_004326
Gene Alias LGS/MGC131591
Gene Symbols BCL9
Host Species Mouse
Immunogen BCL9 (NP_004317, 1036 a.a. ∼ 1135 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 607
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.