Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCL6 co-repressor, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004657 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-657 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-657 Supplier Abnova Corporation Supplier No. H00054880A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant BCOR.

The protein encoded by this gene was identified as an interacting corepressor of BCL6, a POZ/zinc finger transcription repressor that is required for germinal center formation and may influence apoptosis. This protein selectively interacts with the POZ domain of BCL6, but not with eight other POZ proteins. Specific class I and II histone deacetylases (HDACs) have been shown to interact with this protein, which suggests a possible link between the two classes of HDACs. Several transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: RRFRKRPEPSSDYDLSPAKQEPKPFDRLQQLLPASQSTQLPCSSSPQETTQSRPMPPEARRLIVNKNAGETLLQRAARLGYEEVVLYCLENKICDVNHRD

Specifications

Antigen BCL6 co-repressor
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant BCOR.
Formulation 50% glycerol
Gene BCOR
Gene Accession No. NM_017745
Gene Alias ANOP2/FLJ20285/FLJ38041/KIAA1575/MAA2/MCOPS2/MGC131961/MGC71031
Gene Symbols BCOR
Host Species Mouse
Immunogen BCOR (NP_060215, 1361 a.a. ∼ 1460 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 54880
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.