Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCR, Mouse, Clone: 2E5, Abnova™

Catalog No. 89016297 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-297 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-297 Supplier Abnova Corporation Supplier No. H00000613M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BCR.

A reciprocal translocation between chromosomes 22 and 9 produces the Philadelphia chromosome, which is often found in patients with chronic myelogenous leukemia. The chromosome 22 breakpoint for this translocation is located within the BCR gene. The translocation produces a fusion protein which is encoded by sequence from both BCR and ABL, the gene at the chromosome 9 breakpoint. Although the BCR-ABL fusion protein has been extensively studied, the function of the normal BCR gene product is not clear. The protein has serine/threonine kinase activity and is a GTPase-activating protein for p21rac. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: FHHERGLVKVNDKEVSDRISSLGSQAMQMERKKSQHGAGSSVGDASRPPYRGRSSESSCGVDGDYEDAELNPRFLKDNLIDANGGSRPPWPPLEYQPYQ

Specifications

Antigen BCR
Applications ELISA, Immunofluorescence, In situ PLA, Western Blot
Classification Monoclonal
Clone 2E5
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BCR.
Formulation PBS with no preservative; pH 7.4
Gene BCR
Gene Accession No. NM_004327
Gene Alias ALL/BCR-ABL1/BCR1/CML/D22S11/D22S662/FLJ16453/PHL
Gene Symbols BCR
Host Species Mouse
Immunogen BCR (NP_004318.3, 182 a.a. ∼ 280 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 613
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.