Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BCRP1 Mouse anti-Human, Rat, Clone: 5D3, MilliporeSigma™

Catalog No. MAB4155MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4155MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4155MI Supplier MilliporeSigma Supplier No. MAB4155
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect BCRP1 using this Anti-BCRP1 Antibody, clone 5D3 validated for use in flow cytometry, immunocytochemistry, immunofluoresence, FUNC and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BCRP1
Applications Flow Cytometry, Functional Assay, Immunocytochemistry, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 5D3
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ liquid in buffer containing PBS, pH 7.4, with 0.1% sodium azide.
Gene Accession No. NM_004827.2
Gene Symbols ABCG2, BCRP, BMDP, BCRP1, MXR, ABC15, ABCP, EST157481, MGC102821, MXR1, MRX, CDw338
Host Species Mouse
Immunogen 3T3 cells expressing human ABCG2
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Rat
Content And Storage 2-8°C for 12 months. Do not freeze.
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.