Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-BDNF Clone: 35928.11,

Catalog No. GF35L100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
GF35L100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. GF35L100UG Supplier MilliporeSigma™ Supplier No. GF35L100UG

Mouse Monoclonal Antibody

Specifically detects BDNF Clone: 35928.11 in Human samples, and it is validated for ELISA, Immunohistochemistry, Neutralization, Immuno Blotting

Neurotrophic factors define a family of related proteins, which are important in the development, maintenance, and regulation of neurons in both the peripheral and central nervous system. In addition to the prototypical nerve growth factor (NGF), several other factors have been identified which interact with a family of tyrosine kinase receptors. TrkA is the kinase signaling receptor for NGF, while TrkB mediates signaling for both brain-derived neurotrophic factor (BDNF) and neurotrophin-4/5 (NT-4/5). TrkC is the primary receptor for neurotrophin-3 (NT-3), although this neurotrophin can also activate TrkA and TrkB receptors under certain conditions. Axotomy-induced cell death is one experimental paradigm that has been used to discriminate between the effects of various neurotrophic factors on the survival of different neuronal populations. BDNF, NT-3, and NT-4/5 act as motor neuron survival factors, whereas NGF has no effect on the survival of spinal cord motor neurons. NGF is neuroprotective for cholinergic neurons in the central nervous system. Retrogradely transported NGF-TrkA has also been shown to signal the CREB transcription factor in sympathetic neurons. A putative new member of this gene family, neurotrophin-6 (NT-6), has also been identified from the fish Xephophorus. Like NGF, NT-6 promotes the survival of sympathetic and sensory neurons, albeit with a lower potency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BDNF
Applications ELISA, Immunoblot, Immunoblot, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 35928.11
Conjugate Unconjugated
Formulation Lyophilized from PBS, 5% trehalose.
Host Species Mouse
Immunogen recombinant protein consisting of amino acids 129-247 of human BDNF
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽20kDa BDNF protein in brain extracts. Clone 35928.11 was selected based on its ability to neutralize the biological activity of human recombinant BDNF.
Target Species Human
Content And Storage −20°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.