Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin Mouse anti-Human, Mouse, Rat, Clone: 4E10.1, MilliporeSigma™

Catalog No. MABC139MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC139MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC139MI Supplier MilliporeSigma Supplier No. MABC139

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

Detect Beclin using this mouse monoclonal antibody, Anti-Beclin Antibody1 and clone 4E10.1 validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin1
Applications Western Blot
Classification Monoclonal
Clone 4E10.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bλ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_003757
Gene Symbols BECN1, GT197
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to the N-terminus of human Beclin1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.