Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin 1 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. AB15417MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB15417MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB15417MI Supplier MilliporeSigma Supplier No. AB15417
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect Beclin 1 using this Anti-Beclin 1 Antibody validated for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin 1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Lyophilized. Reconstitute with 50μL of sterile distilled water. Centrifuge to remove any insoluble material. Contains no preservative.
Gene Symbols BECN1; GT197; Beclin-1; beclin1; ATG6; VPS30
Host Species Rabbit
Immunogen Synthetic peptide corresponding to the C-terminus of human Beclin 1 protein.
Purification Method Serum
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_003766.2
Target Species Human
Content And Storage Lyophilized: −20-−70°C for up to 12 months. Reconstituted: −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.

Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.