Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beclin 2 Antibody, HRP, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB11060982H
Description
Beclin 2 Polyclonal specifically detects Beclin 2 in Human samples. It is validated for Western Blot.Specifications
Beclin 2 | |
Polyclonal | |
Western Blot | |
A8MW95 | |
BECN2 | |
A synthetic peptide made to an N-terminal region (within residues 1-100) of the human Beclin 2 protein. [Uniprot # A8MW95] | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
IgG |
Western Blot | |
HRP | |
PBS with No Preservative | |
BECN1 L1, BECN1L1, BECN1P1, BECN1P1 beclin 1, autophagy related, pseudogene 1 | |
Rabbit | |
50 kDa | |
0.1 mL | |
Apoptosis, Autophagy, Cancer, Tumor Suppressors | |
441925 | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction