Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beclin Mouse, Unlabeled, Clone: 20, BD

Catalog No. BDB612112 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612112 50 μg
BDB612113 150 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB612112 Supplier BD Biosciences Supplier No. 612112
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Autophagy is the bulk degradation of cellular proteins through the autophagosomic-lysosomal pathway. This process is important for normal cell growth, and may be disrupted in tumor cells. Beclin is a Bcl-2 binding protein that is homologous to the yeast autophagy gene, apg6/vps30. The structure of beclin includes a Bcl-2 binding coiled-coil region, and a leucine-rich nuclear export signal (NES). Beclin protein colocalizes with intracytoplasmic organelles and nuclei in normal COS7 and MCF7 cells. However, inhibition of CRM1-dependent nuclear export leads to beclin localization primarily in the nucleus. Mutation of the NES domain of beclin also prevents nuclear export, as well as suppresses beclin-mediated nutrient deprivation-induced autophagy. In addition, these beclin mutants can not inhibit in vitro clonigenicity and in vivo tumorigenicity of MCF7 cells. Beclin interaction with Bcl-2 may be involved with host viral defense, since overexpression of beclin inhibits Sindbis virus replication and expression of beclin lacking the Bcl-2 binding domain has no antiviral effects. Thus, beclin may be an important component of complexes involved in autophagy.

Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Beclin
Applications Western Blot
Classification Monoclonal
Clone 20
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human Beclin aa. 171-291
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.