Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bestrophin Mouse anti-Human, Monkey, Pig, MilliporeSigma™

Catalog No. MAB5466MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5466MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5466MI Supplier MilliporeSigma Supplier No. MAB5466
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Bestrophin using this Anti-Bestrophin Antibody validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bestrophin
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Conjugate Unconjugated
Formulation Purified immunoglobulin. Liquid in PBS containing 0.08% sodium azide.
Gene Accession No. NM_004183.2
Gene Symbols BEST1, BEST, VMD2, bestrophin, Bestrophin-1, TU15B, BMD
Host Species Mouse
Immunogen Recombinant human bestrophin protein.
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Monkey, Pig
Content And Storage −20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.