Learn More
Description
Specifically detects beta Actin Clone: in Human, Mouse, Rat samples, and it is validated for Immunocytochemistry, Western Blotting
Specifications
Specifications
| Antigen | beta Actin |
| Applications | Immunocytochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal antibody in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. |
| Gene Accession No. | P60709 |
| Gene Symbols | ACTB |
| Host Species | Rabbit |
| Immunogen | C-terminally KLH-conjugated linear peptide corresponding to human beta-actin N-terminal sequence with arginylation modification. |
| Purification Method | Affinity Purified |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.