Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Amyloid 1-42 Rabbit anti-Chicken, Human, Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB5078PMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5078PMI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5078PMI Supplier MilliporeSigma Supplier No. AB5078P
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Beta-Amyloid 1-42 Antibody is an antibody against Beta-Amyloid 1-42 for use in ELISA, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta-Amyloid 1-42
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in liquid buffer containing PBS with 0.1% BSA.
Gene Symbols APP; PN-II; AD1; ABETA; CTFgamma; APPI; ABPP; A4; CVAP; PN2; PreA4; AAA
Host Species Rabbit
Immunogen A six amino acid peptide sequence from the C-terminal of human beta-amyloid 1-42.
Purification Method Affinity Purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000484.2
Target Species Chicken, Human, Mouse
Content And Storage −20°C for up to 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.