Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Arrestin 1 Mouse anti-Human, Clone: 3H11.2, MilliporeSigma™

Catalog No. MAB2609MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB2609MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB2609MI Supplier MilliporeSigma Supplier No. MAB2609
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-beta Arrestin 1 Antibody, clone 3H11.2 detects level of beta Arrestin 1 and has been published and validated for use in western blotting and immunofluoresence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Arrestin 1
Applications Immunofluorescence, Western Blot
Classification Monoclonal
Clone 3H11.2
Conjugate Unconjugated
Formulation Purified in PBS with 0.05% NaN3.
Gene Accession No. NP_004032.2
Gene Symbols ARRESTIN, S-AG, S-arrestin
Host Species Mouse
Immunogen KLH conjugated synthetic peptide corresponding to amino acids 357-372 of human beta arrestin 1.
Purification Method Ammonium sulfate precipitation
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.