Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Rat beta-Catenin Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody has been used in 4 publications

$200.50 - $538.00

Specifications

Antigen beta-Catenin
Clone 196621
Concentration LYOPH
Dilution Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Applications Western Blot, Immunohistochemistry
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1329SP
GREENER_CHOICE
View Documents
R&D Systems
MAB1329SP
25 μg
Each for $200.50
Only null left
Add to Cart
 
MAB1329050
GREENER_CHOICE
View Documents
R&D Systems
MAB1329050
50 μg
Each for $315.50
Only null left
Add to Cart
 
MAB1329200
GREENER_CHOICE
View Documents
R&D Systems
MAB1329200
200 μg
Each for $538.00
Only null left
Add to Cart
 
Description

Description

beta-Catenin Monoclonal specifically detects beta-Catenin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

beta-Catenin
LYOPH
Western Blot, Immunohistochemistry
Unconjugated
Mouse
Human, Mouse, Rat
P35222
1499
E. coli-derived recombinant human beta-Catenin Ala2-Leu781 Accession # P35222
Primary
Reconstitute at 0.5 mg/mL in sterile PBS.
Detects rh beta-Catenin and endogenous human, mouse and rat beta-Catenin in Western blots.
196621
Western Blot 2 ug/mL, Immunohistochemistry 8-25 ug/mL
Monoclonal
Purified
RUO
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
bCatenin, beta 1 (88kD), beta-catenin, catenin (cadherin-associated protein), beta 1, 88kDa, catenin beta-1, CTNNB, CTNNB1, DKFZp686D02253, FLJ25606, FLJ37923
CTNNB1
IgG2b
Protein A or G purified from hybridoma culture supernatant
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.