Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta Galactosidase Chicken anti-Bacteria, Polyclonal, MilliporeSigma™

Catalog No. AB3403IMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB3403IMI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB3403IMI Supplier MilliporeSigma Supplier No. AB3403I
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Beta Galactosidase Antibody, E. coli is a highly specific chicken polyclonal antibody and that targets β-Galactosidase and has been tested in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta Galactosidase
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified chicken polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols GLB1; ELNR1
Host Species Chicken
Immunogen Recombinant protein corresponding to E. Coli Beta Galactosidase.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_752394
Target Species Bacteria
Content And Storage 2-8°C for 12 months
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.