Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta III Tubulin, Cy3 Conjugate Rabbit anti-Human, Mouse, Rat, Cy3, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB15708C3
Description
This Anti-Beta III Tubulin, Cy3 Conjugate Antibody is validated for use in immunocytochemistry for the detection of Beta III Tubulin.
Specifications
Beta III Tubulin | |
Polyclonal | |
Purified rabbit polyclonal conjugated to Cy3 in PBS with 0.1% sodium azide and 15mg/mL BSA. | |
Rabbit | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat |
Immunocytochemistry | |
Cyanine3 | |
TUBB3; TUBB4; MC1R; beta-4 | |
Synthetic linear peptide corresponding to human Beta III Tubulin. | |
100 μL | |
NP_006077 | |
2-8°C for 12 months |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction