Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta III Tubulin Chicken anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB9354MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB9354MI 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB9354MI Supplier MilliporeSigma Supplier No. AB9354
Add to Cart
Add to Cart

Chicken Polyclonal Antibody

Anti-Beta III Tubulin Antibody detects level of Beta III Tubulin and has been published and validated for use in immunocytochemistry, immunohistochemistry and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Beta III Tubulin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Affinity purified polyclonal chicken IgY liquid in PBS, pH 7.2 with 0.02% sodium azide.
Four different antibodies were combined to make this product
Gene Symbols TUBB3; TUBB4; beta-4; MC1R
Host Species Chicken
Immunogen Synthetic peptides from human/rat Beta III Tubulin.
Purification Method Affinity Purified
Quantity 500 μL
Primary or Secondary Primary
Gene ID (Entrez) NM_006086.2
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquotes for up to 6 months
Isotype IgY
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.