Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-beta-Interferon, Polyclonal

Catalog No. 40729920KU Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 kU
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
40-729-920KU 20 kU Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-729-920KU Supplier MilliporeSigma™ Supplier No. 40729920KU
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects β-Interferon Clone: in Human samples, and it is validated for Neutralization

TRUSTED_SUSTAINABILITY

Specifications

Antigen β-Interferon
Applications Neutralization
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum diluted in PBS.
Host Species Rabbit
Immunogen recombinant, human, beta;-interferon
Quantity 20 kU
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽20kDa IFN-β protein. Note: 1 KU = 1000 neutralization units.
Target Species Human
Content And Storage −20°C
Form Diluted
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.