Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TGR5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA523168
Description
BG37 is a 330 amino acid novel enzyme with seven transmembrane domains. It belongs to the G protein-coupled receptor (GPCR) superfamily and functions as a cell surface receptor for bile acids. Cells expressing BG37 when treated with bile acids induce the production of intracellular cAMP, activate the MAP kinase signaling pathway, leads to the internalization of the receptor and regulates proglucagon gene expression in enteroendocrine cells. The receptor is implicated in the suppression of macrophage functions and regulation of energy homeostasis by bile acids that can be targeted to improve metabolic control. It is known to be ubiquitously expressed in most of the tissues.
Specifications
TGR5 | |
Polyclonal | |
GPBAR1 | |
BG37, GPCR19, GPR131, M-BAR, TGR5, G-protein coupled bile acid receptor 1, G-protein coupled bile acid receptor BG37, G-protein coupled receptor GPCR19, hBG37, hGPCR19, membrane bile acid receptor, membrane-type receptor for bile acids | |
Rabbit | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
Q8TDU6, Q80SS6 | |
GPBAR1 | |
Synthetic peptide (207 CRDEPSALARALTWRQARAQA 227) of BG37/TGR5 protein. | |
RUO | |
151306, 227289 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction