Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-BHLHE40/DEC1, Polyclonal

Catalog No. ABN1737MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
ABN1737MI 100 μL Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABN1737MI Supplier MilliporeSigma™ Supplier No. ABN1737
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Specifically detects BHLHE40/DEC1 Clone: in Human samples, and it is validated for Western Blotting

Class E basic helix-loop-helix protein 40 (UniProt O14503; also known as Basic helix-loop-helix domain containing class B2, Class B basic helix-loop-helix protein 2, Differentially expressed in chondrocytes 1, Differentially expressed in chondrocytes protein 1, Differentiated embryo chondrocyte expressed gene 1, Enhancer-of-split and hairy-related protein 2, SHARP-2, Stimulated by retinoic acid gene 13 protein) is encoded by the BHLHE40 (also known as BHLHB2, DEC1, HLHB2, SHARP2, STRA13) gene (Gene ID 8553) in human. DEC1 (differentially expressed in chondrocytes 1) and DEC2 are E-box-binding transcription factors that exhibit a circadian expression pattern. Both proteins act to repress the Clock/Bmal1-activated E-box promoters via direct E-box binding rather than Bmal1 interaction. In addition, DEC1 is shown to negatively regulate the expression of DEC2. Markedly increased DEC1 expression is observed in colon carcinomas, whereas the opposite trend is seen with DEC2. Likewise cellular DEC1 over-expression results in a proportional decrease in DEC2 level.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BHLHE40/DEC1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Rabbit polyclonal serum with 0.05% sodium azide.
Gene Accession No. O14503
Gene Symbols BHLHE40, BHLHB2, DEC1, SHARP2, STRA13
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to human BHLHE40/DEC1.
Purification Method Unpurified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_003661
Target Species Human
Content And Storage −20°C for one year
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.