Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mouse BID cleavage (59/60) site specific Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB10002MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB10002MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB10002MI Supplier MilliporeSigma Supplier No. AB10002
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This Anti-mouse BID cleavage (59/60) site specific Antibody is validated for use in western blotting for the detection of mouse BID cleavage (59/60) site specifimmunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BID cleavage (59/60) site specific
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Dulbecco's phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol with 1.0mg/mL BSA (IgG, protease free) as a carrier, 0.05% sodium azide.
Gene Symbols BID
Host Species Rabbit
Immunogen The antiserum was produced against a chemically synthesized peptide derived from the 15 kDa caspase-8 cleavage product of mouse BID.
Quantity 100 μL
Primary or Secondary Primary
Target Species Mouse
Content And Storage −20°C in undiluted aliquotes
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.