Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bid Mouse anti-Human, Unlabeled, Clone: 7, BD

Catalog No. BDB611528 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB611528 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB611528 Supplier BD Biosciences Supplier No. 611528
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Members of the Bcl-2 protein family function to inhibit (Bcl-2, Bcl-XL, Mcl-1, A1) or promote (Bax, Bak, Bcl-Ls, Bad) apoptosis. Bid is a Bcl-2 family death agonist that heterodimerizes with either agonists (Bax) or antagonists (Bcl-2). Bid contains a centrally located BH3 domain that allows interaction with the BH1 domain of Bax and Bcl-2. However, Bid does not contain other domains commonly found in Bcl-2 family members, such as BH1, BH2, or BH4. In addition, Bid does not contain a C-terminal hydrophobic region that is characteristic of membrane-bound Bcl-2 family members. Singlet oxygen-induced apoptosis in human leukemia cells and Fas-induced apoptosis involve caspase-8 cleavage of Bid, which produces a 15 kDa C-terminal fragment and a 6.5 kDa N-terminal fragment. The C-terminal fragment translocates to the mitochondria and promotes the release of cytochrome C through a mechanism that might involve destabilization of the mitochondrial membrane. Thus, the proapoptotic role of Bid may involve mitochondrial membrane destabilization, as well as multiple protein-protein interactions.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Bid
Applications Western Blot
Classification Monoclonal
Clone 7
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse Bid aa. 50-62
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.