Learn More
Description
Specifications
Specifications
| Antigen | Bim |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Immunoaffinity Purified immunoglobulin in 20mM sodium phosphate, 250mM NaCl, pH. 7.6, with 0.1% sodium azide as a preservative. |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide corresponding to amino acids 22-40 of human origin (O'Conneor et al. 1998). The sequence is identical to that of mouse and is different by one amino acid from that of rat. |
| Purification Method | Immunoaffinity purified |
| Quantity | 100 μg |
| Primary or Secondary | Primary |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.