Learn More
BimEL (pS69) human / (pS65) rat Rabbit anti-Human, Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | BimEL (pS69) human / (pS65) rat |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Dulbecco's phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3 (+/- 0.1), 50% glycerol, with 1.0mg/mL BSA (IgG, protease free) as a carrier. 0.05% sodium azide |
| Gene Symbols | BCL2L11; BIM; BOD; BimEL; BIM-beta6; BimL; BIM-beta7; BAM; BIM-alpha6 |
| Host Species | Rabbit |
| Immunogen | The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of rat BimEL that contains serine 65 (serine 69 in the human sequence). The sequence is conserved in human and mouse. |
| Purification Method | Affinity Purified |
| Quantity | 100 μL |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.