Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bin1 Mouse anti-Human, Chicken, Mouse, Clone: 99D, MilliporeSigma™

Catalog No. 05449CMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-449-CMI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-449-CMI Supplier MilliporeSigma Supplier No. 05449C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Bin1, clone 99D (Ascites Free) Antibody is validated for use in Western Blotting and Immunohistochemistry and Immunoprecipitation and Flow Cytometry and Immunofluorescence for the detection of Bin1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Bin1
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 99D
Concentration 1mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_004296.1
Gene Symbols BIN1, AMPHL
Host Species Mouse
Immunogen Linear peptide corresponding to human Bin1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Chicken, Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.