Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Biotin Antibody, FITC - F(ab), Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP197372
Description
Biotin Polyclonal specifically detects Biotin in Non-species specific samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
Biotin | |
Polyclonal | |
Fluorescein | |
Lyophilized from 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free with 0.01% Sodium Azide | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Reconstitute with 1.0 ml deionized water (or equivalent) | |
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot, Immunoassay | |
LYOPH | |
Western Blot 1:100-1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/Immunofluorescence 1:500-1:2500, Immunohistochemistry-Paraffin 1:20-1:50, Dot Blot, Knockout Validated | |
Vitamin B4 | |
Biotin conjugated to Keyhole Limpet Hemocyanin (b-KLH) | |
1 mg | |
Cancer, Cellular Signaling, Epitope Tags, metabolism | |
Biotin | |
Non-species specific | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction