Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BiP/GRP78 Mouse, Unlabeled, Clone: 40, BD

Catalog No. BDB610979 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610979 150 μg
BDB610978 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610979 Supplier BD Biosciences Supplier No. 610979
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Synthesis of nascent proteins occurs at sites on the endoplasmic reticulum (ER) called translocons. Translocon proteins form a pore in the membrane that allow passage of the newly synthesized protein from the ribosome into the ER lumen. As the nascent protein enters the lumen, it is bound by BiP ( bi nding p rotein), the major chaperone of the ER. This protein is identical to the 78kDa glucose regulated protein, GRP78. BiP binds short hydrophobic sequences of the emerging peptide and prevents denaturation or nonspecific aggregation. Hydrolysis of ATP by BiP results in the release of the nascent protein which quickly assumes its proper conformation. However, if folding is incorrect, BiP again binds the protein and prevents its exit from the ER. In addition, BiP binding is thought to enhance the movement of secretory polypeptides across the ER membrane, but it is not required for protein translocation. It is 60% identical to Hsp70 and is similarly increased by conditions that produce incorrectly folded proteins. Thus, BiP is a chaperone of the ER lumen that binds misfolded or unassembled secretory proteins and ensures proper movement of proteins from the ER to the Golgi apparatus.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BiP/GRP78
Applications Western Blot
Classification Monoclonal
Clone 40
Concentration 250μg/mL
Conjugate Unconjugated
Description GRP78
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human BiP/GRP78 aa. 525-628
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG2a
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.