Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC2/IAP-2 Rat anti-Mouse, Human, Clone: 1E1-1-10, MilliporeSigma™

Catalog No. MABC535MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC535MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC535MI Supplier MilliporeSigma Supplier No. MABC535
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Anti-BIRC2/IAP-2 antibody, clone 1E1-1-10 is an antibody against BIRC2/IAP-2 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BIRC2/IAP-2
Applications Western Blot
Classification Monoclonal
Clone 1E1-1-10
Conjugate Unconjugated
Formulation Purified rat monoclonal IgG2aκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_031491
Gene Symbols Birc2, Birc3, lap2
Host Species Rat
Immunogen Linear peptide corresponding to the BIR 2 repeat region of mouse BIRC2/IAP-2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.