Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIRC4 (M01J), Mouse anti-Human, Clone: 2B3, Abnova™

Catalog No. 89016169 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-169 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-169 Supplier Abnova Corporation Supplier No. H00000331M01J

Mouse monoclonal antibody raised against a full-length recombinant BIRC4.

The protein encoded by this gene is a member of a family of proteins which inhibit apoptosis through binding to tumor necrosis factor receptor-associated factors TRAF1 and TRAF2. This protein inhibits apoptosis induced by menadione, a potent inducer of free radicals, and ICE. It also inhibits at least two members of the caspase family of cell-death proteases, caspase-3 and caspase-7. [provided by RefSeq

Sequence: GFLYTGEGDTVRCFSCHAAVDRWQYGDSAVGRHRKVSPNCRFINGFYLENSATQSTNSGIQNGQYKVENYLGSRDHFALDRPSETHADYLLRTGQVVDIS

Specifications

Antigen BIRC4
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2B3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant BIRC4.
Formulation PBS with no preservative; pH 7.4
Gene XIAP
Gene Accession No. BC032729
Gene Alias API3/BIRC4/ILP1/MIHA/XLP2
Gene Symbols XIAP
Host Species Mouse
Immunogen BIRC4 (AAH32729, 51 a.a. ∼ 150 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 331
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.