Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

baculoviral IAP repeat-containing 8, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89008444 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-008-444 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-008-444 Supplier Abnova Corporation Supplier No. H00112401B01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human BIRC8 protein.

Sequence: MTGYEARLITFGTWMYSVNKEQLARAGFYAIGQEDKVQCFHCGGGLANWKPKEDPWEQHAKWYPGCKYLLEEKGHEYINNIHLTRSLEGALVQTTKKTPSLTKRISDTIFPNPMLQEAIRMGFDFKDVKKIMEERIQTSGSNYKTLGVLVADLVSAQKDTTENELNQTSLQREISPEEPLRRLQEEKLCKICMDRHIAVVFIPCGHLVTCKQCAEAVDRCPMCSMVIDFKQRVFMS

Specifications

Antigen baculoviral IAP repeat-containing 8
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human BIRC8 protein.
Formulation No additive
Gene BIRC8
Gene Accession No. BC039318
Gene Alias ILP-2/ILP2/hILP2
Gene Symbols BIRC8
Host Species Mouse
Immunogen BIRC8 (AAH39318, 1 a.a. ∼ 236 a.a) full-length human protein.
Quantity 50 μL
Regulatory Status RUO
Research Discipline Apoptosis
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 112401
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.