Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-BKV VP1, Clone: 1C7, Abnova™
Mouse Monoclonal Antibody
Supplier: Abnova Corporation MAB3204M03
Specifications
BKV VP1 | |
Monoclonal | |
Unconjugated | |
In 1x PBS, pH 7.4 | |
VP1 | |
Mouse | |
100 μg | |
Primary | |
Virus | |
Purified |
ELISA, Western Blot | |
1C7 | |
Mouse monoclonal antibody raised against a full-length recombinant BKV VP1. | |
NC_001538 | |
BKPyVgp4 | |
BKV VP1 (YP_717939.1, 1 a.a. ∼ 369 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
RUO | |
1489515 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction