Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-BKV VP1, Clone: 6D1, Abnova™

Catalog No. 89300379 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-300-379 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-300-379 Supplier Abnova Corporation Supplier No. MAB3204M11
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifications

Antigen BKV VP1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6D1
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full-length recombinant BKV VP1.
Formulation In 1x PBS, pH 7.4
Gene Accession No. NC_001538
Gene Alias VP1
Gene Symbols BKPyVgp4
Host Species Mouse
Immunogen BKV VP1 (YP_717939.1, 1 a.a. ∼ 369 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1489515
Target Species Virus
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.