Learn More
Blimp-1 Rat anti-Mouse, Alexa Fluor 647, Clone: 5E7., BD
Rat Monoclonal Antibody
Supplier: BD Biosciences 563643
Description
The 5E7 monoclonal antibody specifically binds to mouse B lymphocyte-induced maturation protein 1 (Blimp-1). Blimp-1 is a 98 kDa zinc finger-containing protein that is encoded by the Prdm1 gene and functions as a transcriptional repressor. Blimp-1 is essential for regulating the terminal differentiation of activated B cells into plasma cells. In this process, Blimp-1 suppresses genes involved in B cell proliferation, such as Myc and Bcl6. It controls other genes, such as Xbp1, that are required for upregulating the plasma cell’s protein synthesis machinery which is essential for antibody production and secretion. Blimp-1 is an important regulator in other cell types as well. It functions in the regulation of T cell activation and homeostasis and NK cell maturation. Blimp-1 is expressed in activated T cells and can function to inhibit IL-2 production while enhancing the differentiation and expression of several molecules produced by effector T cells.

Specifications
Blimp-1 | |
Monoclonal | |
0.2mg/mL | |
Prdm1; PR domain zinc finger protein 1; Blimp1; PRDI-BF1; ZNFPR1A1 | |
Mouse Blimp-1 Recombinant Protein | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
5E7. | |
Alexa Fluor 647 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.