Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BLK, Mouse, Clone: 7A12, Abnova™
Mouse monoclonal antibody raised against a partial recombinant BLK.
Supplier: Abnova Corporation H00000640M02
Description
Sequence: MGLVSSKKPDKEKPIKEKDKGQWSPLKVSAQDKDAPPLPPLVVFNHLTPPPPDEHLDEDKHFVVALYDYTAMNDRDLQMLKGEKLQVLKGSpecifications
BLK | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC007371 | |
BLK | |
BLK (AAH07371, 1 a.a. ∼ 90 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
ELISA, Western Blot | |
7A12 | |
Mouse monoclonal antibody raised against a partial recombinant BLK. | |
BLK | |
MGC10442 | |
Mouse | |
Affinity chromatography | |
RUO | |
640 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction