Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLNK Mouse, Alexa Fluor 647, Clone: 2B11, BD

Catalog No. BDB558692 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558692 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558692 Supplier BD Biosciences Supplier No. 558692

Mouse Monoclonal Antibody

B cell activation is initiated by crosslinking the B cell receptor, which leads to activation of non-receptor protein tyrosine kinases (PTK), including Btk, Syk, and three Src kinases, Fyn, Lyn, and Blk. Activated PTKs then phosphorylate multiple cellular proteins involved in B lymphocyte signaling. Syk is responsible for the tyrosine phosphorylation of B cell l i nk er protein (BLNK), a member of the SLP-76 family of adapter proteins. Phosphorylation of human BLNK at tyrosines 84, 178, and 189 (Y84, Y178, and Y189) creates docking sites for PLC g

The 2B11 monoclonal antibody recognizes BLNK, regardless of phosphorylation status. A fusion protein representing amino acids 4-205 of human BLNK was used as the immunogen. BLNK is expressed as two phosphoproteins migrating at 68 and 70 kDa in SDS/PAGE that represent alternatively spliced forms of human BLNK.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BLNK
Applications Flow Cytometry
Classification Monoclonal
Clone 2B11
Conjugate Alexa Fluor 647
Description SLP-65, BASH, BCA
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human N-terminal BLNK Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.