Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLNK Mouse, Unlabeled, Clone: 2B11, BD

Catalog No. BDB559930 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB559930 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB559930 Supplier BD Biosciences Supplier No. 559930
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

B cell activation is initiated by crosslinking the B cell receptor, which leads to activation of non-receptor protein tyrosine kinases (PTK), including Btk, Syk, and three Src kinases, Fyn, Lyn, and Blk. Activated PTKs then phosphorylate multiple cellular proteins involved in B lymphocyte signaling. Syk is responsible for the tyrosine phosphorylation of B cell l i nk er protein (BLNK), a member of the SLP-76 family of adapter proteins. Phosphorylation of human BLNK at tyrosines 84, 178, and 189 (Y84, Y178, and Y189) creates docking sites for PLC g

The 2B11 monoclonal antibody recognizes BLNK, regardless of phosphorylation status. A fusion protein representing amino acids 4-205 of human BLNK was used as the immunogen. BLNK is expressed as two phosphoproteins migrating at 68 and 70 kDa in SDS/PAGE that represent alternatively spliced forms of human BLNK.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BLNK
Applications Western Blot
Classification Monoclonal
Clone 2B11
Concentration 0.5mg/mL
Conjugate Unconjugated
Description SLP-65, BASH, BCA
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human N-terminal BLNK Recombinant Protein
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store undiluted at 4°C
Isotype IgG2a κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.