Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BLTR-1 Mouse anti-Human, PE, Clone: 203/14F11, BD

Catalog No. BDB552836 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB552836 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB552836 Supplier BD Biosciences Supplier No. 552836

Mouse Monoclonal Antibody

The monoclonal antibody 203/14F11 reacts against human leukotriene B4 receptor 1 (BLTR-1). BLTR-1 is a seven-transmembrane, G-protein-coupled receptor. The receptor plays an important role in pro-inflammatory responses. Leukotrienne B4 receptors are distributed in various hematopoietic cells including monocytes, granulocytes, and lymphocytes. Leukotriene B4 (LTB4), a productof arachidonic acid metabolism, is the ligand for BLTR-1. The immunogen used to generate 203/14F11 hybridoma was a human BLTR-1-transfected cell line.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BLTR-1
Applications Flow Cytometry
Classification Monoclonal
Clone 203/14F11
Conjugate PE
Description hBLTR-1
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Human BLTR-1-transfected cell line
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.