Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Blys (BAFF) Mouse anti-Human, Unlabeled, Clone: H32-406, BD

Catalog No. BDB558289 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558289 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558289 Supplier BD Biosciences Supplier No. 558289

Mouse Monoclonal Antibody

Antibody H32-406 reacts with B-lymphocyte stimulator protein (Blys), also known as BAFF (B cell activating factor) and is a member of the TNF ligand superfamily. Members of the TNF receptor and ligand superfamily play a critical role in regulating immune responses. Blys is a type II membrane protein expressed on cells of myeloid lineage including monocytes, dendritic cells, and activated T cells. Blys exists in both a membrane-bound and soluble form. In its soluble form, Blys is a potent B cellgrowth factor in costimulation assays, suggesting its biological role in monocyte-driven B cell activation. Blys has also been linked to B cell hyperplasia and autoimmunity.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Blys (BAFF)
Applications ELISA
Classification Monoclonal
Clone H32-406
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.