Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BM28 Mouse, Unlabeled, Clone: 46, BD

Catalog No. BDB610701 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
150 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB610701 150 μg
BDB610700 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB610701 Supplier BD Biosciences Supplier No. 610701
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

BM28 is a phosphoprotein that migrates at 125kDa in SDS PAGE with a hyperphosphorylated form migrating as a slightly slower band. The ratio of the phosphorylation states alters with the phases of cell cycle - in M, the “fast” phosphorylated BM28 is the major form found, while in G1 phase, the “slow”, hyperphosphorylated BM28 predominates. The cellular localization of BM28 is also cell cycle dependent. In G1, most of the BM28 is chromatin bound, whereas, by M phase, the BM28 is still nuclear, but not associated with the DNA. BM28 is necessary for both entry into S phase and cell division as determined by microinjection inhibition experiments. Microinjection of a BM28 antibody into synchronised cells in G1 phase inhibits DNA replication. When injected during S phase or later, no effect on DNA replication is seen, but cell division is inhibited.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen BM28
Applications Western Blot
Classification Monoclonal
Clone 46
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Purification Method Affinity Purified
Quantity 150 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Canine, Chicken, Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.