Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Rabbit anti-Human, Mouse, Rat, Opossum, Bovine, Primate, Monkey, Polyclonal, MilliporeSigma™

Catalog No. AB2298MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2298MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2298MI Supplier MilliporeSigma Supplier No. AB2298

Rabbit Polyclonal Antibody

This Anti-BMAL1 Antibody is validated for use in western blotting for the detection of BMAL1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMAL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal antibody in 0.1M Tris-Glycine (pH7.4) 150mM NaCl with 0.05% NaN3.
Host Species Rabbit
Immunogen KLH conjugated synthetic linear peptide corresponding to the internal domain of BMAL1.
Purification Method Antigen affinity purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_001075390.1
Target Species Bovine, Human, Monkey, Mouse, Opossum, Primate, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.