Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMAL1 Guinea Pig anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB2204MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB2204MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB2204MI Supplier MilliporeSigma Supplier No. AB2204
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

Anti-BMAL1 Antibody is an antibody against BMAL1 for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMAL1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Serum with 0.05% NaN3
Gene Symbols BMAL1; BMAL1c; JAP3; MGC47515; MOP3; PASD3; TIC
Host Species Guinea Pig
Immunogen Recombinant Protein
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_031515
Target Species Human, Mouse, Rat
Content And Storage −20°C in undiluted aliquotes for 12 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.