Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMI1 Mouse anti-Human, FITC, Clone: AF27, MilliporeSigma™

Catalog No. FCMAB149FMI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FCMAB149FMI 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FCMAB149FMI Supplier MilliporeSigma Supplier No. FCMAB149F

Mouse Monoclonal Antibody

Milli-Mark™ Anti-BMI1 -FITC Antibody, clone AF27 detects level of BMI1 and has been published and validated for use in flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMI1
Applications Flow Cytometry
Classification Monoclonal
Clone AF27
Conjugate FITC
Formulation Purified mouse monoclonal IgG1κ conjugated to FITC in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_005171
Gene Symbols MGC12685, PCGF4, RNF51, flvi-2/bmi-1
Host Species Mouse
Immunogen MBP-tagged recombinant protein corresponding to full length human BMI1.
Purification Method Protein G purfied
Quantity 100 Tests
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.