Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMP8B, Mouse, Clone: 6D6, Abnova™

Catalog No. 89016347 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-347 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-347 Supplier Abnova Corporation Supplier No. H00000656M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a partial recombinant BMP8B.

The bone morphogenetic proteins (BMPs) are a family of secreted signaling molecules that can induce ectopic bone growth. Many BMPs are part of the transforming growth factor-beta (TGFB) superfamily. BMPs were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. Based on its expression early in embryogenesis, the BMP encoded by this gene has a proposed role in early development. In addition, the fact that this BMP is closely related to BMP5 and BMP7 has led to speculation of possible bone inductive activity. [provided by RefSeq

Sequence: KKSNELPQANRLPGIFDDVHGSHGRQVCRRHELYVSFQDLGWLDWVIAPQGYSAYYCEGECSFPLDSCMNATNHAILQSLVHLMMPDAV

Specifications

Antigen BMP8B
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6D6
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a partial recombinant BMP8B.
Formulation ascites with no preservative
Gene BMP8B
Gene Accession No. NM_001720
Gene Alias BMP8/MGC131757/OP2
Gene Symbols BMP8B
Host Species Mouse
Immunogen BMP8B (NP_001711, 274 a.a. ∼ 362 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 656
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.