Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-1A Mouse anti-Human, Mouse, Clone: 4B7B2, MilliporeSigma™

Catalog No. MABS419MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS419MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS419MI Supplier MilliporeSigma Supplier No. MABS419
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-BMPR-1A, clone 4B7B2 Antibody is validated for use in western blotting, immunohistochemistry, flow cytometry and ELISA for the detection of BMPR-1A.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BMPR-1A
Applications Flow Cytometry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 4B7B2
Conjugate Unconjugated
Formulation Mouse monoclonal IgG1 ascitic fluid containing up to 0.1% sodium azide.
Gene Symbols BMPR1A, ACVRLK3, ALK3
Host Species Mouse
Immunogen Purified recombinant fragment of human BMPR1A expressed in E. Coli
Purification Method Unpurified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage −20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.