Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BMPR-II Mouse, Unlabeled, Clone: 18, BD

Catalog No. BDB612292 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB612292 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB612292 Supplier BD Biosciences Supplier No. 612292
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The transforming growth factor β (TGFβ)/activin/BMP family of growth factor plays a diverse and important role in growth, development, and differentiation. The receptors for this famiy of proteins are type I and II ser/thr kinase receptors. Bone morphogenetic protein receptors (BMPRs) include two type I receptors, BMPR-IA and BMPR-IB, and a type II receptor, BMPR-II. BMPR-II is a widely expressed receptor, with high mRNA expression during development in many tissues. BMPR-II transfected COS-1 cells show binding to BMP-7 (osteogenic protein-1) and BMP-4, and co-transfection with the type I receptor, ActR-I, leads to transcriptional activation in response to BMP-7. In addition, extracellular matrix proteins may regulate the expression of BMPR-II during angiogenesis, since fibrillar type I collagen induces BMPR-II expression in endothelial cells. In familial primary pulmonary hypertension, the BMPR-II gene is mutated in a manner that may disrupt ligand binding, kinase activity, and heteromeric dimer formation. Thus, BMPR-II may be an important mediator of BMP effects during growth and differentiation of endothelial cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen BMPR-II
Applications Western Blot
Classification Monoclonal
Clone 18
Concentration 250μg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Mouse BMPR-II aa. 803-996
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at -20°C.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.